Learn More
Alkali Scientific nUVaClean™ UV Pipette Carousel
SDP
Catalog No. 502335743
Encompass_Preferred
ProPette™ nUVaClean is the first and only pipette rack that not only organizes and protects six pipettes, but also destroys greater than 99% of unwanted contamination. The new nUVaClean uses a germicidal UV-C lamp. A high-efficiency reflector/concentrator completely bathes pipette shafts in 254nm UV light. This exposure destroys unwanted microbiological contaminants and prevents cross-contamination in PCR and other sensitive procedures. Just place single-channel pipettes of any range or brand in the nUVaClean and press the “start” button to begin the auto-decon cycle. After 28 minutes the nUVaClean will power down automatically, delivering up to 6 pipettes that are safe, clean and ready for use.
Catalog No. 50-233-5743 Supplier Alkali Scientific Supplier No. P3590
ProPette™ nUVaClean is the first and only pipette rack that not only organizes and protects six pipettes, but also destroys greater than 99% of unwanted contamination. The new nUVaClean uses a germicidal UV-C lamp. A high-efficiency reflector/concentrator completely bathes pipette shafts in 254nm UV light. This exposure destroys unwanted microbiological contaminants and prevents cross-contamination in PCR and other sensitive procedures. Just place single-channel pipettes of any range or brand in the nUVaClean and press the “start” button to begin the auto-decon cycle. After 28 minutes the nUVaClean will power down automatically, delivering up to 6 pipettes that are safe, clean and ready for use.