Learn More
Research Products International Corp nUVaClean UV Pipette Carousel
SDP
Catalog No. 502114083
Encompass
The ProPette nUVaClean is the first and only pipette rack that organizes and protects six pipettes, as well as destroys >99% surface contamination. The new nUVaClean uses a germicidal UV-C lamp with a high-efficiency reflector/ concentrator to completely bathe pipette shafts in 254nm light with 360° UV exposure. UV-C exposure reduces and destroys biological contaminants, mycoplasma, bacteria, fungi, viruses, and DNA up to 99%. This treatment destroys unwanted microbiological contaminants and prevents cross contamination in PCR and other sensitive procedures.
Easy to use - place up to six single channel pipettes in the nUVaClean Carousel and press the “start” button. One step decontamination cycle begins immediately, and after 28 minutes, the unit powers down automatically, delivering pipettes that are safe, clean and ready for use.
Catalog No. 50-211-4083 Supplier Research Products International Corp Supplier No. 247195
The ProPette nUVaClean is the first and only pipette rack that organizes and protects six pipettes, as well as destroys >99% surface contamination. The new nUVaClean uses a germicidal UV-C lamp with a high-efficiency reflector/ concentrator to completely bathe pipette shafts in 254nm light with 360° UV exposure. UV-C exposure reduces and destroys biological contaminants, mycoplasma, bacteria, fungi, viruses, and DNA up to 99%. This treatment destroys unwanted microbiological contaminants and prevents cross contamination in PCR and other sensitive procedures.
Easy to use - place up to six single channel pipettes in the nUVaClean Carousel and press the “start” button. One step decontamination cycle begins immediately, and after 28 minutes, the unit powers down automatically, delivering pipettes that are safe, clean and ready for use.