Learn More
Medchemexpress LLC HY-U00119 1mg Medchemexpress, Nuvenzepine CAS:96487-37-5 Purity:>98%
SDP
Catalog No. 501934420
Encompass_Preferred
Medchemexpress, HY-U00119 1mg Nuvenzepine CAS:96487-37-5 Nuvenzepine is an mAChR antagonist previously in phase I clinical trials for the treatment of gastrospasm. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4420 Supplier Medchemexpress LLC Supplier No. HYU001191MG
Medchemexpress, HY-U00119 1mg Nuvenzepine CAS:96487-37-5 Nuvenzepine is an mAChR antagonist previously in phase I clinical trials for the treatment of gastrospasm. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.