Learn More
Bio Rad Laboratories Nuvia™ HR-S Media, 100 ml #1560513
Catalog No. NC3655249
Encompass
100 ml, strong cation exchange chromatography media, 50 µm particle size
Catalog No. NC3655249 Supplier Bio Rad Laboratories Supplier No. 1560513
100 ml, strong cation exchange chromatography media, 50 µm particle size