Learn More
Bio Rad Laboratories Nuvia™ HR-S Media, 500 ml #1560515
Catalog No. NC3655298
Encompass
500 ml, strong cation exchange chromatography media, 50 µm particle size
Catalog No. NC3655298 Supplier Bio Rad Laboratories Supplier No. 1560515
500 ml, strong cation exchange chromatography media, 50 µm particle size