Learn More
MSC NUVO A/F SFTY GLASSES
Catalog No. 5000787756
Encompass
These half-framed safety glasses feature clear, anti-fog and scratch-resistant polycarbonate lenses. Designed for eye protection, they offer a comfortable fit with adjustable temples and meet ANSI Z87.1+ and CSA Z94.3 safety standards.
  • Anti-fog and scratch-resistant lens coating
  • Half-framed wrap around design
  • Clear polycarbonate lenses
  • Gray frame and temple color
  • Universal size with adjustable temples
  • UV protection
  • Meets CSA Z94.3 and ANSI Z87.1+ standards
  • Features soft nose bridge, integral brow bar, and side shields
Catalog No. 50-007-87756 Supplier MSC Supplier No. 54132444
These half-framed safety glasses feature clear, anti-fog and scratch-resistant polycarbonate lenses. Designed for eye protection, they offer a comfortable fit with adjustable temples and meet ANSI Z87.1+ and CSA Z94.3 safety standards.
  • Anti-fog and scratch-resistant lens coating
  • Half-framed wrap around design
  • Clear polycarbonate lenses
  • Gray frame and temple color
  • Universal size with adjustable temples
  • UV protection
  • Meets CSA Z94.3 and ANSI Z87.1+ standards
  • Features soft nose bridge, integral brow bar, and side shields