Learn More
Medchemexpress LLC HY-114384 5mg , NV-5138 CAS:2095886-80-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1143845MG
Encompass_Preferred
HY-114384 5mg NV-5138 CAS: 2095886-80-7 NV-5138, a leucine analog, is the first selective and orally active brain mTORC1 activator, binding to Sestrin2. NV-5138 is used for antidepressant studies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9636