Learn More
Medchemexpress LLC HY-114384B 5mg , NV-5138 (hydrochloride) CAS: Purity:>98%
SDP
Catalog No. 502259319
Encompass_Preferred
HY-114384B 5mg NV-5138 (hydrochloride) CAS: NV-5138 hydrochloride, a leucine analog, is the first selective and orally active brain mTORC1 activator, binding to Sestrin2. NV-5138 hydrochloride is used for antidepressant studies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9319 Supplier Medchemexpress LLC Supplier No. HY114384B5MG
HY-114384B 5mg NV-5138 (hydrochloride) CAS: NV-5138 hydrochloride, a leucine analog, is the first selective and orally active brain mTORC1 activator, binding to Sestrin2. NV-5138 hydrochloride is used for antidepressant studies. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.