Learn More
Sino Biological Norovirus GII.4 VP1 Protein (His & MBP Tag)
Catalog No. 502993310
Encompass_Preferred
A DNA sequence encoding the Norovirus GII.4 (Norovirus Hu/GII.4/Sydney/NSW0514/2012/AU) VP1 (4OOS_A) (Thr1-Met309) was expressed with a N-terminal polyhistidine tag followed by a MBP tag.
Catalog No. 50-299-3310 Supplier Sino Biological Supplier No. 41002V10E100
A DNA sequence encoding the Norovirus GII.4 (Norovirus Hu/GII.4/Sydney/NSW0514/2012/AU) VP1 (4OOS_A) (Thr1-Met309) was expressed with a N-terminal polyhistidine tag followed by a MBP tag.