Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific NV RAM 4636 120V cool

Catalog No. NC1558308 Shop All Thermo Scientific Products
Encompass_Preferred

For Thermo Scientific Brand Centrifuges model SL 40R - 120V 60Hz

Catalog No. NC1558308 Supplier Thermo Scientific Supplier No. 70904636
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For Thermo Scientific Brand Centrifuges model SL 40R - 120V 60Hz

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.