Learn More
Cayman Chemical Nevirapine-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1051418-95-1, 1 mg
SDP
Catalog No. 502939707
Encompass_Preferred
An internal standard for the quantification of nevirapine by GC- or LC-MS
Catalog No. 50-293-9707 Supplier Cayman Chemical Supplier No. 280931 MG
An internal standard for the quantification of nevirapine by GC- or LC-MS