Learn More
Cayman Chemical nvIrapIn-d4 5mg
SDP
Supplier:  Cayman Chemical 280935 MG
Encompass_Preferred
An internal standard for the quantification of nevirapine by GC- or LC-MS
Catalog No. 50-294-0806