Learn More
Aviva Systems Biology Corp NVL Antibody - N-terminal region (ARP61610_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61610_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NVL. It was validated on Western Blot. . Alternate names: -ApplicationWB. Species reactivity: Cow, Horse, Human, Pig, Rat
Catalog No. 50-287-419