Learn More
Medchemexpress LLC HY-12214A 10mg , NVP-2 CAS:1263373-43-8 Purity:>98%
SDP
Catalog No. 502259986
Encompass_Preferred
HY-12214A 10mg NVP-2 CAS: 1263373-43-8 NVP-2 is a potent and selective ATP-competitive cyclin dependent kinase 9 (CDK9) probe, inhibits CDK9/CycT activity with an IC50 of 0.514 nM. NVP-2 displays inhibitory effcts on CDK1/CycB, CDK2/CycA and CDK16/CycY kinases with IC50 values of 0.584 µM, 0.706 µM, and 0.605 µM, respectively. NVP-2 induces cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9986 Supplier Medchemexpress LLC Supplier No. HY12214A10MG
HY-12214A 10mg NVP-2 CAS: 1263373-43-8 NVP-2 is a potent and selective ATP-competitive cyclin dependent kinase 9 (CDK9) probe, inhibits CDK9/CycT activity with an IC50 of 0.514 nM. NVP-2 displays inhibitory effcts on CDK1/CycB, CDK2/CycA and CDK16/CycY kinases with IC50 values of 0.584 µM, 0.706 µM, and 0.605 µM, respectively. NVP-2 induces cell apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.