Learn More
TARGETMOL CHEMICALS INC NVP-2 5MG
SDP
Catalog No. 502577341
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NVP-2 is an effective and selective ATP-competitive cyclin-dependent kinase 9 (CDK9) probe. NVP-2 induces cell apoptosis. NVP-2 inhibits CDK9/CycT activity (IC50 0.514 nM). NVP-2 shows inhibitory effcts on CDK1/CycB CDK2/CycA and CDK16/CycY kinases (IC50 0.584 uM 0.706 uM and 0.605 uM respectively). purity: 99%
Catalog No. 50-257-7341 Supplier TARGETMOL CHEMICALS INC Supplier No. T163635MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NVP-2 is an effective and selective ATP-competitive cyclin-dependent kinase 9 (CDK9) probe. NVP-2 induces cell apoptosis. NVP-2 inhibits CDK9/CycT activity (IC50 0.514 nM). NVP-2 shows inhibitory effcts on CDK1/CycB CDK2/CycA and CDK16/CycY kinases (IC50 0.584 uM 0.706 uM and 0.605 uM respectively). purity: 99%