Learn More
Echelon Biosciences Research Labs NVP-231 (Ceramide Kinase Inhibitor) 1 mg
SDP
Supplier:  Echelon Biosciences Research Labs B00401MG
Encompass
NVP-231 is a potent, specific, and reversible Ceramide kinase (CerK) inhibitor (IC50= 12 nM in vitro) which competitively inhibits binding of ceramide. CerK is a key enzyme for controlling ceramide levels. This item is non returnable
Catalog No. 50-822-49