Learn More
Medchemexpress LLC HY-13945 10mg Medchemexpress, NVP 231 CAS:362003-83-6 Purity:>98%
SDP
Catalog No. 502081536
Encompass_Preferred
Medchemexpress, HY-13945 10mg NVP 231 CAS:362003-83-6 NVP 231 is a potent, specific, and reversible ceramide kinase (CerK) inhibitor(IC50=12 nM) that competitively inhibits binding of ceramide to CerK. NVP 231 induces cell apoptosis by increasing DNA fragmentation and caspase-3 and caspase-9 cleavage. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1536 Supplier Medchemexpress LLC Supplier No. HY1394510MG
Medchemexpress, HY-13945 10mg NVP 231 CAS:362003-83-6 NVP 231 is a potent, specific, and reversible ceramide kinase (CerK) inhibitor(IC50=12 nM) that competitively inhibits binding of ceramide to CerK. NVP 231 induces cell apoptosis by increasing DNA fragmentation and caspase-3 and caspase-9 cleavage. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.