Learn More
Medchemexpress LLC NVP-ADW742 | 475488-23-4 | MFCD16038913 | 99.3% | 453.58 g/mol | C28H31N5O | 10 MG
SDP
Catalog No. 5000248402
Encompass_Preferred
NVP-ADW742 is a small-molecule, ATP-competitive inhibitor of the insulin-like growth factor 1 receptor (IGF-1R) used in research on cancer signaling pathways. It has demonstrated submicromolar potency and is supplied as a high-purity research reagent.
  • Selective IGF-1R inhibition with submicromolar potency.
  • High purity suitable for research applications.
  • Available in multiple small-scale package sizes, including 10 mg.
  • Powder stable under frozen storage for long-term use.
  • Applicable for in vitro and in vivo experimental studies on IGF-1R signaling.
Catalog No. 50-002-48402 Supplier Medchemexpress LLC Supplier No. HY1025210MG
NVP-ADW742 is a small-molecule, ATP-competitive inhibitor of the insulin-like growth factor 1 receptor (IGF-1R) used in research on cancer signaling pathways. It has demonstrated submicromolar potency and is supplied as a high-purity research reagent.
  • Selective IGF-1R inhibition with submicromolar potency.
  • High purity suitable for research applications.
  • Available in multiple small-scale package sizes, including 10 mg.
  • Powder stable under frozen storage for long-term use.
  • Applicable for in vitro and in vivo experimental studies on IGF-1R signaling.