Learn More
TARGETMOL CHEMICALS INC NVP-AEW541 10MG
SDP
Catalog No. 502579584
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NVP-AEW541 (AEW541) a potent inhibitor of IGF-1R(IC50=150 nM) and InsR(IC50=140 nM) exhibits excellent efficiency and specificity for IGF-1R in a cell-based assay. purity: 99%
Catalog No. 50-257-9584 Supplier TARGETMOL CHEMICALS INC Supplier No. T608010MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NVP-AEW541 (AEW541) a potent inhibitor of IGF-1R(IC50=150 nM) and InsR(IC50=140 nM) exhibits excellent efficiency and specificity for IGF-1R in a cell-based assay. purity: 99%