Learn More
Selleck Chemical LLC NVP-AEW541 2mg 475489-16-8 AEW541
SDP
Catalog No. NC0485434
Encompass_Preferred
NVP-AEW541 is a potent inhibitor of IGF-1R/InsR with IC50 of 150 nM/140 nM in cell-free assays, greater potency and selectivity for IGF-1R in a cell-based assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0485434 Supplier Selleck Chemical LLC Supplier No. S10342MG
NVP-AEW541 is a potent inhibitor of IGF-1R/InsR with IC50 of 150 nM/140 nM in cell-free assays, greater potency and selectivity for IGF-1R in a cell-based assay. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.