Learn More
Medchemexpress LLC HY-100394 2mg Medchemexpress, NVP-BAW2881 CAS:861875-60-7 Purity:>98%
SDP
Catalog No. 501931807
Encompass_Preferred
Medchemexpress, HY-100394 2mg NVP-BAW2881 CAS:861875-60-7 NVP-BAW2881 (BAW2881) is a potent and selective VEGFR2 inhibitor with an IC 50 of 4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1807 Supplier Medchemexpress LLC Supplier No. HY1003942MG
Medchemexpress, HY-100394 2mg NVP-BAW2881 CAS:861875-60-7 NVP-BAW2881 (BAW2881) is a potent and selective VEGFR2 inhibitor with an IC 50 of 4 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.