Learn More
Medchemexpress LLC HY-13258A 5mg Medchemexpress, NVP-BHG712 CAS:940310-85-0 Purity:>98%
SDP
Catalog No. 501969840
Encompass_Preferred
Medchemexpress, HY-13258A 5mg NVP-BHG712 CAS:940310-85-0 NVP-BHG712 is an oral active EphB4 kinase autophosphorylation inhibitor, with IC50 values of 3.3 nM and 3.0 nM for EphA2 and EphB4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9840 Supplier Medchemexpress LLC Supplier No. HY13258A5MG
Medchemexpress, HY-13258A 5mg NVP-BHG712 CAS:940310-85-0 NVP-BHG712 is an oral active EphB4 kinase autophosphorylation inhibitor, with IC50 values of 3.3 nM and 3.0 nM for EphA2 and EphB4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.