Learn More
Selleck Chemical LLC NVP-BHG712 50mg 940310-85-0
SDP
Supplier:  Selleck Chemical LLC S220250MG
Encompass_Preferred
NVP-BHG712 is a specific EphB4 inhibitor with ED50 of 25 nM that discriminates between VEGFR and EphB4 inhibition; also shows activity against c-Raf, c-Src and c-Abl with IC50 of 0.395 ?M, 1.266 ?M and 1.667 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-556-1