Learn More
Medchemexpress LLC HY-13258 5mg Medchemexpress, NVP-BHG712 isomer CAS:2245892-85-5 Purity:>98%
SDP
Catalog No. 502081318
Encompass_Preferred
Medchemexpress, HY-13258 5mg NVP-BHG712 isomer CAS:2245892-85-5 NVP-BHG712 isomer, a regioisomer of NVP-BHG712, shows conserved non-bonded binding to EPHA2 and EPHB4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1318 Supplier Medchemexpress LLC Supplier No. HY132585MG
Medchemexpress, HY-13258 5mg NVP-BHG712 isomer CAS:2245892-85-5 NVP-BHG712 isomer, a regioisomer of NVP-BHG712, shows conserved non-bonded binding to EPHA2 and EPHB4. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.