Learn More
Medchemexpress LLC HY-70063 500mg , NVP-BKM120 Buparlisib;BKM120 CAS:944396-07-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY70063500MG
Encompass_Preferred
Medchemexpress, HY-70063 500mg NVP-BKM120 Buparlisib;BKM120 CAS:944396-07-0 Formula:C18H21F3N6O2 IC50: 52 nM (p110α), 166 nM (p110β), 116 nM (p110δ), 262 nM (p110γ) Purity:98% NVP-BKM120 is a pan-class I PI3K inhibitor, with IC 50 of 52 nM/166 nM/116 nM/262 nM for p110α / p110β / p110δ / p110γ , respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2336