Learn More
Selleck Chemical LLC NVP-BSK805 2HCl S2686-1g
SDP
Catalog No. 503112531
Encompass_Preferred
NVP-BSK805 2HCl is a potent and selective ATP-competitive JAK2 inhibitor with IC50 of 0 5 nM 20-fold selectivity towards JAK1 JAK3 and TYK2
Catalog No. 50-311-2531 Supplier Selleck Chemical LLC Supplier No. S26861G
NVP-BSK805 2HCl is a potent and selective ATP-competitive JAK2 inhibitor with IC50 of 0 5 nM 20-fold selectivity towards JAK1 JAK3 and TYK2