Learn More
Medchemexpress LLC NVP-BSK805 dihydrochloride | 1942919-79-0 | 99.0% | 563.47 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14722A
Encompass_Preferred
NVP-BSK805 dihydrochloride is an ATP-competitive inhibitor primarily targeting JAK2, with high selectivity over other JAK family members like JAK1, JAK3, and TYK2. It effectively suppresses the growth of JAK2V617F-bearing acute myeloid leukemia cell lines and blocks STAT5 phosphorylation. This compound also enhances the sensitivity of drug-resistant cancer cells to certain treatments by improving P-gp inhibitory activity.
  • Potent ATP-competitive JAK2 inhibitor
  • Selective inhibition over JAK1, JAK3, and TYK2
  • Suppresses JAK2V617F-bearing acute myeloid leukemia cell growth
  • Blocks STAT5 phosphorylation
  • Improves P-gp inhibitory activity
  • Increases sensitization of drug-resistant cancer cells to treatments
Catalog No. 50-003-63129