Learn More
Medchemexpress LLC HY-15954 5mg Medchemexpress, NVP-CGM097 CAS:1313363-54-0 Purity:>98%
SDP
Catalog No. 502081847
Encompass_Preferred
Medchemexpress, HY-15954 5mg NVP-CGM097 CAS:1313363-54-0 NVP-CGM097 is a potent and selective MDM2 inhibitor with IC50 of 1.7±0.1 nM for hMDM2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1847 Supplier Medchemexpress LLC Supplier No. HY159545MG
Medchemexpress, HY-15954 5mg NVP-CGM097 CAS:1313363-54-0 NVP-CGM097 is a potent and selective MDM2 inhibitor with IC50 of 1.7±0.1 nM for hMDM2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.