Learn More
Medchemexpress LLC HY-15954A 2mg Medchemexpress, NVP-CGM097 (stereoisomer) CAS: Purity:>98%
SDP
Catalog No. 501932759
Encompass_Preferred
Medchemexpress, HY-15954A 2mg NVP-CGM097 (stereoisomer) CAS: NVP-CGM097 (stereoisomer) is a stereoisomer of NVP-CGM097, with no special bioactivity. NVP-CGM097 is a potent and selective MDM2 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2759 Supplier Medchemexpress LLC Supplier No. HY15954A2MG
Medchemexpress, HY-15954A 2mg NVP-CGM097 (stereoisomer) CAS: NVP-CGM097 (stereoisomer) is a stereoisomer of NVP-CGM097, with no special bioactivity. NVP-CGM097 is a potent and selective MDM2 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.