Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC NVP-CGM097 sulfate | 1313367-56-4 | MFCD31382179 | 98.8% | 757.34 g/mol | C38H49ClN4O8S | 10 MG
SDP

Catalog No. 5000288105
Encompass_Preferred

NVP-CGM097 sulfate is a potent, selective small-molecule inhibitor of MDM2 supplied as the sulfate salt for research applications. It binds human MDM2 with high potency (IC50 1.7 ± 0.1 nM) and is provided at high reported purity for biochemical and cellular studies.

  • Potent MDM2 inhibitor with IC50 1.7 ± 0.1 nM for hMDM2.
  • Provided as the sulfate salt.
  • Reported purity of 98.82%.
  • Molecular weight 757.34 g/mol and formula C38H49ClN4O8S.
  • Available in small-scale quantities suitable for laboratory research.

Catalog No. 50-002-88105 Supplier Medchemexpress LLC Supplier No. HY15954B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NVP-CGM097 sulfate is a potent, selective small-molecule inhibitor of MDM2 supplied as the sulfate salt for research applications. It binds human MDM2 with high potency (IC50 1.7 ± 0.1 nM) and is provided at high reported purity for biochemical and cellular studies.

  • Potent MDM2 inhibitor with IC50 1.7 ± 0.1 nM for hMDM2.
  • Provided as the sulfate salt.
  • Reported purity of 98.82%.
  • Molecular weight 757.34 g/mol and formula C38H49ClN4O8S.
  • Available in small-scale quantities suitable for laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.