Learn More
Abcam NVP DPP 728, DPP-IV inhibitor, 5MG
Catalog No. 502706298
Encompass_Preferred
MW 371.3 Da, Purity >98%. Potent, selective, reversible dipeptidyl peptidase-IV (DPP-IV) inhibitor (Ki = 11 nM). Antidiabetic agent in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6298 Supplier Abcam Supplier No. AB1421715MG
MW 371.3 Da, Purity >98%. Potent, selective, reversible dipeptidyl peptidase-IV (DPP-IV) inhibitor (Ki = 11 nM). Antidiabetic agent in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement