Learn More
TARGETMOL CHEMICALS INC NVP-TAE 226 5MG
SDP
Catalog No. 502878578
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. NVP-TAE 226 (TAE226) is a potent FAK inhibitor (IC50 5.5 nM), exhibiting even greater efficacy against Pyk2 (IC50 3.5 nM), and is 10- to 100-fold less effective against IGF-1R, InsR, c-Met, and ALK. Purity 98.07%
Catalog No. 50-287-8578 Supplier TARGETMOL CHEMICALS INC Supplier No. T19185MG
Also available in 1 mL, 1 mg, 2 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. NVP-TAE 226 (TAE226) is a potent FAK inhibitor (IC50 5.5 nM), exhibiting even greater efficacy against Pyk2 (IC50 3.5 nM), and is 10- to 100-fold less effective against IGF-1R, InsR, c-Met, and ALK. Purity 98.07%