Learn More
Medchemexpress LLC Nvp-tae 226 | 761437-28-9 | MFCD12031516 | 99.8% | 468.94 g·mol⁻¹ | C23H25ClN6O3 | 5 MG
SDP
Catalog No. 5000262613
Encompass_Preferred
NVP-TAE 226 is an ATP-competitive small-molecule inhibitor of focal adhesion kinase (FAK) and insulin-like growth factor 1 receptor (IGF-1R), used in cellular and preclinical cancer research to probe signaling pathways and evaluate effects on proliferation, migration, and survival.
  • Potent inhibition of FAK and Pyk2 with low-nanomolar IC50s.
  • Also inhibits IGF-1R and insulin receptor, enabling study of connected signaling networks.
  • Suitable for in vitro assays and preclinical efficacy models.
  • Available as solid and solution formulations for flexible dosing.
  • High purity appropriate for research applications.
  • Stable under recommended storage conditions for prolonged shelf life.
Catalog No. 50-002-62613 Supplier Medchemexpress LLC Supplier No. HY132035MG
NVP-TAE 226 is an ATP-competitive small-molecule inhibitor of focal adhesion kinase (FAK) and insulin-like growth factor 1 receptor (IGF-1R), used in cellular and preclinical cancer research to probe signaling pathways and evaluate effects on proliferation, migration, and survival.
  • Potent inhibition of FAK and Pyk2 with low-nanomolar IC50s.
  • Also inhibits IGF-1R and insulin receptor, enabling study of connected signaling networks.
  • Suitable for in vitro assays and preclinical efficacy models.
  • Available as solid and solution formulations for flexible dosing.
  • High purity appropriate for research applications.
  • Stable under recommended storage conditions for prolonged shelf life.