Learn More
Medchemexpress LLC Nvp-tae 226 | 761437-28-9 | MFCD12031516 | 99.8% | C23H25ClN6O3 | 10MG
SDP
Catalog No. 5000260980
Encompass_Preferred
NVP-TAE 226 is a potent, ATP-competitive small-molecule inhibitor of focal adhesion kinase (FAK) and insulin-like growth factor 1 receptor (IGF-1R), used in preclinical oncology and signal-transduction research. It exhibits activity across multiple kinases and has demonstrated antitumor effects in vitro and in vivo.
  • Potent FAK inhibition (IC50 5.5 nM).
  • Dual activity against IGF-1R and Pyk2 (IC50 140 nM and 3.5 nM).
  • Additional activity against InsR, c-Met, KDR, and Flt3.
  • High purity suitable for research (~99.8%).
  • Stable as powder under recommended storage conditions.
Catalog No. 50-002-60980 Supplier Medchemexpress LLC Supplier No. HY1320310MG
NVP-TAE 226 is a potent, ATP-competitive small-molecule inhibitor of focal adhesion kinase (FAK) and insulin-like growth factor 1 receptor (IGF-1R), used in preclinical oncology and signal-transduction research. It exhibits activity across multiple kinases and has demonstrated antitumor effects in vitro and in vivo.
  • Potent FAK inhibition (IC50 5.5 nM).
  • Dual activity against IGF-1R and Pyk2 (IC50 140 nM and 3.5 nM).
  • Additional activity against InsR, c-Met, KDR, and Flt3.
  • High purity suitable for research (~99.8%).
  • Stable as powder under recommended storage conditions.