Learn More
TARGETMOL CHEMICALS INC NVP-TAE 684 10MG
SDP
Catalog No. 502577188
Encompass_Preferred
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NVP-TAE 684 (TAE684) is a excellently effective and specific ALK inhibitor(IC50=3 nM). purity: 99%
Catalog No. 50-257-7188 Supplier TARGETMOL CHEMICALS INC Supplier No. T225110MG
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. NVP-TAE 684 (TAE684) is a excellently effective and specific ALK inhibitor(IC50=3 nM). purity: 99%