Learn More
Medchemexpress LLC NVP-TAE 684 | 761439-42-3 | 99.7% | 614.20 g/mol | C30H40ClN7O3S | 100 MG
SDP
Catalog No. 5000249134
Encompass_Preferred
NVP-TAE 684 is a potent, selective anaplastic lymphoma kinase (ALK) inhibitor used in research to block proliferation of ALCL-derived and ALK-dependent cell lines. It shows low-nanomolar activity and is supplied as a high-purity small molecule for in vitro studies and compound screening.
  • Potent ALK inhibition with IC50 in the low-nanomolar range (≈2-10 nM).
  • Selective activity against ALK-dependent and ALCL-derived cell lines.
  • Suitable for in vitro research and compound screening applications.
  • Available as solid and DMSO solution formats and multiple pack sizes.
  • High purity suitable for research use (>99.7%).
Catalog No. 50-002-49134 Supplier Medchemexpress LLC Supplier No. HY10192100MG
NVP-TAE 684 is a potent, selective anaplastic lymphoma kinase (ALK) inhibitor used in research to block proliferation of ALCL-derived and ALK-dependent cell lines. It shows low-nanomolar activity and is supplied as a high-purity small molecule for in vitro studies and compound screening.
  • Potent ALK inhibition with IC50 in the low-nanomolar range (≈2-10 nM).
  • Selective activity against ALK-dependent and ALCL-derived cell lines.
  • Suitable for in vitro research and compound screening applications.
  • Available as solid and DMSO solution formats and multiple pack sizes.
  • High purity suitable for research use (>99.7%).