Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nvp-TAE 684 | 761439-42-3 | MFCD11977634 | 99.7% | 614.20 | C30H40ClN7O3S | 1 ML
SDP

Catalog No. 5000249135
Encompass_Preferred

NVP-TAE 684 is a potent, selective small-molecule inhibitor of anaplastic lymphoma kinase (ALK) used in preclinical research to block the growth of ALCL-derived and ALK-dependent cell lines (reported IC50 2-10 nM). It is provided as both solid material and as a ready-to-use 10 mM solution in DMSO, and is characterized for purity suitable for in vitro studies.

  • Potent ALK inhibition with low nanomolar IC50 values.
  • Selective activity against ALK-dependent cell lines.
  • Available as a 10 mM solution in DMSO for ready-to-use experiments.
  • High purity (99.7%) suitable for research applications.
  • Molecular weight 614.20 Da and formula C30H40ClN7O3S.
  • Offered in multiple solid pack sizes for flexible dosing and storage.
  • Documentation available including data sheet and SDS.

Catalog No. 50-002-49135 Supplier Medchemexpress LLC Supplier No. HY1019210MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NVP-TAE 684 is a potent, selective small-molecule inhibitor of anaplastic lymphoma kinase (ALK) used in preclinical research to block the growth of ALCL-derived and ALK-dependent cell lines (reported IC50 2-10 nM). It is provided as both solid material and as a ready-to-use 10 mM solution in DMSO, and is characterized for purity suitable for in vitro studies.

  • Potent ALK inhibition with low nanomolar IC50 values.
  • Selective activity against ALK-dependent cell lines.
  • Available as a 10 mM solution in DMSO for ready-to-use experiments.
  • High purity (99.7%) suitable for research applications.
  • Molecular weight 614.20 Da and formula C30H40ClN7O3S.
  • Offered in multiple solid pack sizes for flexible dosing and storage.
  • Documentation available including data sheet and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.