Learn More
Medchemexpress LLC NVP-TAE684 | 761439-42-3 | MFCD11977634 | 99.7% | 614.2 g/mol | C30H40ClN7O3S | 10 MG
SDP
Catalog No. 5000246924
Encompass_Preferred
NVP-TAE684 is a potent, selective small-molecule inhibitor of anaplastic lymphoma kinase (ALK) used in biochemical and cellular research. It blocks growth of ALK-dependent and ALCL-derived cell lines with low-nanomolar IC50 values and has been applied in preclinical studies to probe ALK signaling and validate target engagement. Typically supplied as a solid for in vitro experiments.
  • Potent ALK inhibition at low-nanomolar concentrations.
  • Selective activity against ALK-driven cell lines.
  • Suitable for biochemical and cellular assays.
  • Supplied as a solid for research use.
  • Characterized with reported purity and analytical data.
Catalog No. 50-002-46924 Supplier Medchemexpress LLC Supplier No. HY1019210MG
NVP-TAE684 is a potent, selective small-molecule inhibitor of anaplastic lymphoma kinase (ALK) used in biochemical and cellular research. It blocks growth of ALK-dependent and ALCL-derived cell lines with low-nanomolar IC50 values and has been applied in preclinical studies to probe ALK signaling and validate target engagement. Typically supplied as a solid for in vitro experiments.
  • Potent ALK inhibition at low-nanomolar concentrations.
  • Selective activity against ALK-driven cell lines.
  • Suitable for biochemical and cellular assays.
  • Supplied as a solid for research use.
  • Characterized with reported purity and analytical data.