Learn More
Selleck Chemical LLC NVP-TNKS656 2mg 1419949-20-4
SDP
Catalog No. 501364544
Encompass_Preferred
NVP-TNKS656 is a highly potent, selective, and orally active tankyrase inhibitor with IC50 of 6 nM for TNKS2, > 300-fold selectivity against PARP1 and PARP2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4544 Supplier Selleck Chemical LLC Supplier No. S72382MG
Add to Cart
Add to Cart
NVP-TNKS656 is a highly potent, selective, and orally active tankyrase inhibitor with IC50 of 6 nM for TNKS2, > 300-fold selectivity against PARP1 and PARP2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.