Learn More
Medchemexpress LLC Nvp-tnks656 | 1419949-20-4 | MFCD28167762 | >98.0% | 494.58 g/mol | C27H34N4O5 | 5 MG
SDP
Catalog No. 502081545
Encompass_Preferred
NVP-TNKS656 is a potent, selective, and orally active inhibitor of tankyrase 2 (TNKS2) with a reported IC50 of 6 nM and >300-fold selectivity versus PARP1 and PARP2. It is used as a research tool to study TNKS-mediated regulation of Wnt signaling and related cellular pathways, and is supplied in small research-scale quantities suitable for biochemical and cell-based assays.
  • Potent TNKS2 inhibition (IC50 ≈ 6 nM).
  • High selectivity versus PARP1 and PARP2 (>300-fold).
  • Orally active profile for in vivo study applications.
  • Appropriate for biochemical and cell-based assays.
  • Available in small-mass formats for research use.
Catalog No. 50-208-1545 Supplier Medchemexpress LLC Supplier No. HY139905MG
NVP-TNKS656 is a potent, selective, and orally active inhibitor of tankyrase 2 (TNKS2) with a reported IC50 of 6 nM and >300-fold selectivity versus PARP1 and PARP2. It is used as a research tool to study TNKS-mediated regulation of Wnt signaling and related cellular pathways, and is supplied in small research-scale quantities suitable for biochemical and cell-based assays.
  • Potent TNKS2 inhibition (IC50 ≈ 6 nM).
  • High selectivity versus PARP1 and PARP2 (>300-fold).
  • Orally active profile for in vivo study applications.
  • Appropriate for biochemical and cell-based assays.
  • Available in small-mass formats for research use.