Learn More
Medchemexpress LLC HY-50866 10mg Medchemexpress, NVP-AEW541 CAS:475489-16-8 Purity:>98%
SDP
Catalog No. 502029344
Encompass_Preferred
Medchemexpress, HY-50866 10mg NVP-AEW541 CAS:475489-16-8 NVP-AEW541 is a potent inhibitor of IGF-1R with IC50 of 0.15 μM, also inhibits InsR, with IC50 of 0.14 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9344 Supplier Medchemexpress LLC Supplier No. HY5086610MG
Medchemexpress, HY-50866 10mg NVP-AEW541 CAS:475489-16-8 NVP-AEW541 is a potent inhibitor of IGF-1R with IC50 of 0.15 μM, also inhibits InsR, with IC50 of 0.14 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.