Learn More
Apexbio Technology LLC NVR 3-778 1445790-55-5 1mg
SDP
Catalog No. 5000093884
Encompass_Preferred
NVR 3-778 (CAS 1445790-55-5) is a small-molecule modulator targeting the hepatitis B virus (HBV) core protein It is designed to interfere with HBV capsid assembly thereby inhibiting viral genome encapsidation and replication NVR 3-778 exerts its biological activity primarily by disrupting capsid formation and reducing viral replication In in vitro studies NVR 3-778 demonstrates antiviral inhibition with a half-maximal inhibitory concentration (IC50) reported in the low micromolar range Based on these pharmacological properties NVR 3-778 holds research potential in the investigation of HBV biology and the development of alternative therapeutic approaches for chronic hepatitis B infection
Catalog No. 50-000-93884 Supplier Apexbio Technology LLC Supplier No. A99551
NVR 3-778 (CAS 1445790-55-5) is a small-molecule modulator targeting the hepatitis B virus (HBV) core protein It is designed to interfere with HBV capsid assembly thereby inhibiting viral genome encapsidation and replication NVR 3-778 exerts its biological activity primarily by disrupting capsid formation and reducing viral replication In in vitro studies NVR 3-778 demonstrates antiviral inhibition with a half-maximal inhibitory concentration (IC50) reported in the low micromolar range Based on these pharmacological properties NVR 3-778 holds research potential in the investigation of HBV biology and the development of alternative therapeutic approaches for chronic hepatitis B infection