Learn More
Medchemexpress LLC Nvr 3-778 | 1445790-55-5 | 99.9% | 432.4 g/mol | C18H16F4N2O4S | 25MG
SDP
Catalog No. 5000237102
Encompass_Preferred
NVR 3-778 is a small-molecule hepatitis B virus (HBV) capsid assembly modulator (CAM) of the sulfamoylbenzamide (SBA) class. It is orally bioavailable and has demonstrated antiviral activity in preclinical models and early clinical studies. Supplied as a solid research compound, typical specifications include CAS 1445790-55-5, molecular formula C18H16F4N2O4S, and molecular weight ≈432.4 g/mol.
  • First-in-class capsid assembly modulator targeting HBV core protein.
  • Orally bioavailable with demonstrated antiviral activity in vitro and in vivo.
  • Provided as a high-purity solid suitable for laboratory research.
  • Includes molecular identity and physicochemical data for characterization.
  • Recommended cold storage to preserve stability in powder and solution.
Catalog No. 50-002-37102 Supplier Medchemexpress LLC Supplier No. HY12460025MG
NVR 3-778 is a small-molecule hepatitis B virus (HBV) capsid assembly modulator (CAM) of the sulfamoylbenzamide (SBA) class. It is orally bioavailable and has demonstrated antiviral activity in preclinical models and early clinical studies. Supplied as a solid research compound, typical specifications include CAS 1445790-55-5, molecular formula C18H16F4N2O4S, and molecular weight ≈432.4 g/mol.
  • First-in-class capsid assembly modulator targeting HBV core protein.
  • Orally bioavailable with demonstrated antiviral activity in vitro and in vivo.
  • Provided as a high-purity solid suitable for laboratory research.
  • Includes molecular identity and physicochemical data for characterization.
  • Recommended cold storage to preserve stability in powder and solution.