Learn More
Selleck Chemical LLC NVR 3-778
SDP
Supplier:  Selleck Chemical LLC S67575MG
Encompass_Preferred
NVR 3-778 belonging to the SBA (sulfamoylbenzamide) class is a first-in-Class and oral bioavailable HBV CAM (capsid assembly modulator) NVR 3-778 exhibits anti-HBV activity
Catalog No. 50-000-02387