Learn More
TARGETMOL CHEMICALS INC NVR 3-778 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T163665MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. NVR 3-778 is a first-in-class, orally bioavailable HBV CAM belonging to the SBA class, exhibiting anti-HBV activity. Purity 99.74%
Catalog No. 50-287-7555