Learn More
Medchemexpress LLC HY-110374 5mg Medchemexpress, NVS-CECR2-1 CAS:1992047-61-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1103745MG
Encompass_Preferred
Medchemexpress, HY-110374 5mg NVS-CECR2-1 CAS:1992047-61-6 NVS-CECR2-1, a non-BET family Bromodomain (BRD) inhibitor, is a potent and selective cat eye syndrome chromosome region, candidate 2 (CECR2) inhibitor. NVS-CECR2-1 binds to CECR2 BRD with high affinity (IC50=47 nM; KD=80 nM). NVS-CECR2-1 exhibits cytotoxic activity and induces apoptosis against various cancer cells by targeting CECR2 as well as via CECR2-independent mechanism. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0783