Learn More
Medchemexpress LLC NVS-SM2 | 1562333-92-9 | 99.15% | 406.52 | 10 MG
SDP
Catalog No. 5000346754
Encompass_Preferred
NVS-SM2 is a potent, orally active, and brain-penetrant SMN2 splicing enhancer. It enhances U1-pre-mRNA association, promotes exon 7 inclusion, and restores normal survival motor neuron (SMN) protein expression. This compound is suitable for spinal muscular atrophy (SMA) research.
  • Potent, orally active, and brain-penetrant SMN2 splicing enhancer
  • Enhances U1-pre-mRNA association
  • Promotes exon 7 inclusion
  • Restores normal survival motor neuron (SMN) protein expression
  • Suitable for spinal muscular atrophy (SMA) research
Catalog No. 50-003-46754 Supplier Medchemexpress LLC Supplier No. HY11152010MG
NVS-SM2 is a potent, orally active, and brain-penetrant SMN2 splicing enhancer. It enhances U1-pre-mRNA association, promotes exon 7 inclusion, and restores normal survival motor neuron (SMN) protein expression. This compound is suitable for spinal muscular atrophy (SMA) research.
  • Potent, orally active, and brain-penetrant SMN2 splicing enhancer
  • Enhances U1-pre-mRNA association
  • Promotes exon 7 inclusion
  • Restores normal survival motor neuron (SMN) protein expression
  • Suitable for spinal muscular atrophy (SMA) research