Learn More
Medchemexpress LLC HY-114395 5mg Medchemexpress, NVS-ZP7-4 CAS:2349367-89-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1143955MG
Encompass_Preferred
Medchemexpress, HY-114395 5mg NVS-ZP7-4 CAS:2349367-89-9 NVS-ZP7-4 is a Zinc transporter SLC39A7 (ZIP7) inhibitor that is also the first reported chemical tool to probe the impact of modulating ER zinc levels and investigate ZIP7 as a novel druggable node in the Notch pathway. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9342