Learn More
Medchemexpress LLC HY-101597 100mg Medchemexpress, NVX-207 CAS:745020-66-0 Purity:>98%
SDP
Catalog No. 501932182
Encompass_Preferred
Medchemexpress, HY-101597 100mg NVX-207 CAS:745020-66-0 NVX-207 is a derivative of betulinic acid with anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2182 Supplier Medchemexpress LLC Supplier No. HY101597100MG
Medchemexpress, HY-101597 100mg NVX-207 CAS:745020-66-0 NVX-207 is a derivative of betulinic acid with anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.