Learn More
Medchemexpress LLC Nvx-207 | 745020-66-0 | MFCD31382160 | 98.0% | 601.86 g/mol | C36H59NO6 | 250 MG
SDP
Catalog No. 5000248257
Encompass_Preferred
NVX-207 is a small-molecule, betulinic acid-derived anticancer research compound supplied as a solid white to off-white powder. It has a reported purity of 98.0%, molecular formula C36H59NO6, molecular weight 601.86 g/mol, and CAS 745020-66-0. Vendor documentation includes a data sheet, SDS, COA, and spectral data for characterization.
  • Provided as a solid white to off-white powder suitable for research use.
  • High reported purity (98.0%) for reliable experimental results.
  • Well-characterized chemical properties (C36H59NO6; MW 601.86 g/mol).
  • Accompanied by documentation including data sheet, SDS, COA, and spectral data.
  • Betulinic acid-derived compound with reported anticancer activity in cell lines.
Catalog No. 50-002-48257 Supplier Medchemexpress LLC Supplier No. HY101597250MG
NVX-207 is a small-molecule, betulinic acid-derived anticancer research compound supplied as a solid white to off-white powder. It has a reported purity of 98.0%, molecular formula C36H59NO6, molecular weight 601.86 g/mol, and CAS 745020-66-0. Vendor documentation includes a data sheet, SDS, COA, and spectral data for characterization.
  • Provided as a solid white to off-white powder suitable for research use.
  • High reported purity (98.0%) for reliable experimental results.
  • Well-characterized chemical properties (C36H59NO6; MW 601.86 g/mol).
  • Accompanied by documentation including data sheet, SDS, COA, and spectral data.
  • Betulinic acid-derived compound with reported anticancer activity in cell lines.